siRNA Details
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027183.2 | 3 | 1317 | |||
NR_045678.1 | 3 | 1294 | ||||
NR_045679.1 | 3 | 1299 | ||||
GenBank mRNA | AK095170.1 | - | 2 | 1915 | ||
BC047586.1 | - | 3 | 1231 |
