siRNA Details
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027183.2 | 3 | 717 | |||
NR_045678.1 | 3 | 694 | ||||
NR_045679.1 | 3 | 699 | ||||
GenBank mRNA | AK095170.1 | - | 2 | 1315 | ||
AK308536.1 | - | 3 | 694 | |||
BC047586.1 | - | 3 | 631 | |||
BX419382.2 | - | 2 | 476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL528925.3 | - | 3 | 500 | Chr. 5 - 180831039 - 180834871 [+] on Build GRCh38 |
