siRNA Details
Gene Symbol: IGFBP7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034081.1 | 5 | 1190 | |||
GenBank mRNA | BC039519.1 | - | 5 | 1190 | ||
BG189240.1 | - | 5 | 401 | |||
BG425784.1 | - | 4 | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG207311.1 | - | 5 | 401 | Chr. 4 - 57179145 - 57205522 [+] on Build GRCh38 |
