siRNA Details
Gene Symbol: SAP30L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037897.1 | 3 | 312 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SAP30L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037897.1 | 3 | 312 |