siRNA Details
Gene Symbol: DDX11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038927.2 | 1 | 637 | |||
GenBank mRNA | AK096360.1 | - | 1 | 641 |
Gene Symbol: LOC105376678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520903.2 | XP_011519205.1 | 1 | 388 |
