siRNA Details
Gene Symbol: NADK2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC033402.1 | - | 2 | 368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG771949.1 | - | 2 | 373 | Chr. 5 - 36221055 - 36221901 [+] on Build GRCh38 |
