siRNA Details
Gene Symbol: ALDH1L1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046383.1 | 2 | 2301 | |||
GenBank mRNA | AK097508.1 | - | 4 | 2212 | ||
BC039521.1 | - | 2 | 1872 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ALDH1L1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046383.1 | 2 | 2301 | |||
GenBank mRNA | AK097508.1 | - | 4 | 2212 | ||
BC039521.1 | - | 2 | 1872 |