siRNA Details
Gene Symbol: ALDH1L1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046383.1 | 2 | 1084 | |||
GenBank mRNA | AK097508.1 | - | 4 | 995 | ||
BC039521.1 | - | 2 | 655 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM805225.1 | - | 2 | 673 | Chr. 3 - 126180490 - 126209171 [+] on Build GRCh38 |
