siRNA Details
Gene Symbol: LOC101929241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110165.1 | 3 | 751 | |||
GenBank mRNA | BC038465.1 | - | 3 | 697 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI833374.1 | - | 3 | 695 | Chr. 14 - 97458883 - 97463918 [+] on Build GRCh38 |
