siRNA Details
Gene Symbol: STARD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040093.1 | 6 | 1430 | |||
GenBank mRNA | CR749489.1 | - | 6 | 1430 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STARD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040093.1 | 6 | 1430 | |||
GenBank mRNA | CR749489.1 | - | 6 | 1430 |