siRNA Details
Gene Symbol: UBL7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038448.1 | 1 | 626 | |||
NR_038449.1 | 1 | 626 | ||||
NR_120377.1 | 1 | 626 | ||||
GenBank mRNA | AF085896.1 | - | 1 | 531 | ||
BC121819.1 | - | 1 | 290 |
