siRNA Details
Gene Symbol: OIP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026757.1 | 2 | 193 | |||
GenBank mRNA | AK172729.1 | - | 2 | 188 | ||
AY007106.1 | - | 2 | 175 | |||
BC037835.1 | - | 2 | 224 | |||
BX647764.1 | - | 2 | 192 |
