siRNA Details
Gene Symbol: CKMT2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034121.1 | 1 | 213 | |||
NR_034122.1 | 1 | 213 | ||||
NR_034123.1 | 1 | 213 | ||||
GenBank mRNA | AK092460.1 | - | 1 | 188 | ||
AK096014.1 | - | 1 | 141 | |||
AK096680.1 | - | 1 | 203 | |||
DB051602.1 | - | 1 | 213 |
