siRNA Details
Gene Symbol: CKMT2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034121.1 | 3 | 1830 | |||
NR_034122.1 | 4 | 1339 | ||||
NR_034123.1 | 2 | 1164 | ||||
GenBank mRNA | AK092460.1 | - | 4 | 1314 | ||
AK096014.1 | - | 3 | 1757 | |||
AK096680.1 | - | 2 | 1154 |
