siRNA Details
Gene Symbol: PCED1B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026544.1 | 3 | 470 | |||
GenBank mRNA | AK123977.1 | - | 3 | 462 | ||
DA925061.1 | - | 2 | 397 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA938003.1 | - | 3 | 462 | Chr. 12 - 47210326 - 47216435 [-] on Build GRCh38 |
