siRNA Details
Gene Symbol: LOC257396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034107.2 | 1 | 117 | |||
NR_104654.1 | 1 | 117 | ||||
GenBank mRNA | AK097893.1 | - | 1 | 102 | ||
BC041894.1 | - | 1 | 60 | |||
BQ637503.1 | - | 1 | 82 | |||
DA398674.1 | - | 1 | 117 | |||
DR158823.1 | - | 1 | 72 |
