siRNA Details
Gene Symbol: TMEM161B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039993.1 | 4 | 613 | |||
NR_039994.2 | 4 | 571 | ||||
NR_039995.1 | 3 | 491 | ||||
NR_105019.1 | 8 | 992 | ||||
NR_105020.1 | 3 | 555 | ||||
GenBank mRNA | BC030122.2 | - | 4 | 557 | ||
BC047059.1 | - | 8 | 984 | |||
BF672668.1 | - | 4 | 546 | |||
BM719684.1 | - | 4 | 613 | |||
DA317999.1 | - | 3 | 525 |
