siRNA Details
Gene Symbol: CCAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108049.1 | 2 | 728 | |||
GenBank mRNA | AK125310.1 | - | 2 | 548 | ||
HM358356.1 | - | 2 | 326 | |||
JX003870.1 | - | 2 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB011555.1 | - | 2 | 366 | Chr. 8 - 127209226 - 127218906 [-] on Build GRCh38 |
