siRNA Details
Gene Symbol: DPH6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038251.1 | 3 | 710 | |||
GenBank mRNA | BC025664.1 | - | 3 | 710 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI756625.1 | - | 3 | 711 | Chr. 15 - 35546195 - 35858323 [+] on Build GRCh38 |
