siRNA Details
Gene Symbol: LINC01194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033383.1 | 1 | 171 | |||
GenBank mRNA | AY328030.1 | - | 1 | 171 | ||
AY328031.1 | - | 1 | 171 | |||
AY328032.1 | - | 1 | 171 | |||
AY328033.1 | - | 1 | 171 | |||
AY330599.1 | - | 1 | 171 |
