siRNA Details
Gene Symbol: SLIT2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037876.2 | 4 | 588 | |||
GenBank mRNA | BI831389.1 | - | 4 | 580 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLIT2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037876.2 | 4 | 588 | |||
GenBank mRNA | BI831389.1 | - | 4 | 580 |