siRNA Details
Gene Symbol: LINC00616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037866.1 | 11 | 1802 | |||
GenBank mRNA | BC042886.1 | - | 11 | 1802 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037866.1 | 11 | 1802 | |||
GenBank mRNA | BC042886.1 | - | 11 | 1802 |