siRNA Details
Gene Symbol: FGF10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108034.1 | 3 | 671 | |||
GenBank mRNA | BC042062.1 | - | 3 | 671 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM546924.1 | - | 3 | 685 | Chr. 5 - 44388730 - 44413054 [+] on Build GRCh38 |
