siRNA Details
Gene Symbol: SRP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040059.1 | 1 | 91 | |||
NR_040060.1 | 1 | 91 | ||||
NR_040061.1 | 1 | 91 | ||||
NR_040062.1 | 1 | 91 | ||||
GenBank mRNA | AK127696.1 | - | 1 | 89 | ||
BM706250.1 | - | 1 | 63 | |||
BX111264.1 | - | 1 | 65 | |||
EB388563.1 | - | 1 | 15 |
