siRNA Details
Gene Symbol: LOC101928651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109910.1 | 5, 6 | 1085 | |||
GenBank mRNA | BC043229.1 | - | 1, 2 | 53 | ||
BC043261.1 | - | 5, 6 | 1090 | |||
BC043516.1 | - | 1, 2 | 62 |
