siRNA Details
Gene Symbol: OTX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029385.1 | 1 | 206 | |||
GenBank mRNA | AB593038.1 | - | 1 | 158 | ||
AB593039.1 | - | 1 | 68 | |||
AB593040.1 | - | 1 | 230 | |||
AB593041.1 | - | 2 | 328 | |||
BC041486.2 | - | 1 | 205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM799998.1 | - | 1 | 216 | Chr. 14 - 56813183 - 56929380 [+] on Build GRCh38 |

More Information
Other Species Targeted

