siRNA Details
Gene Symbol: NPHP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002811.1 | 3 | 205 | |||
GenBank mRNA | BC036463.1 | - | 4 | 868 | ||
BC039375.1 | - | 3 | 205 | |||
BC048124.1 | - | 3 | 204 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NPHP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002811.1 | 3 | 205 | |||
GenBank mRNA | BC036463.1 | - | 4 | 868 | ||
BC039375.1 | - | 3 | 205 | |||
BC048124.1 | - | 3 | 204 |