siRNA Details
Gene Symbol: NPHP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002811.1 | 4, 5 | 334 | |||
GenBank mRNA | BC039375.1 | - | 4, 5 | 334 | ||
BC048124.1 | - | 4, 5 | 333 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NPHP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002811.1 | 4, 5 | 334 | |||
GenBank mRNA | BC039375.1 | - | 4, 5 | 334 | ||
BC048124.1 | - | 4, 5 | 333 |