siRNA Details
Gene Symbol: LINC01093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039975.1 | 3 | 476 | |||
NR_039976.1 | 3 | 463 | ||||
NR_039977.1 | 2 | 243 | ||||
GenBank mRNA | AK096134.1 | - | 3 | 1996 | ||
AY429594.1 | - | 3 | 476 | |||
AY429595.1 | - | 2 | 243 | |||
CB153796.1 | - | 3 | 453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG568575.1 | - | 3 | 471 | Chr. 4 - 184893901 - 184899454 [-] on Build GRCh38 |
