siRNA Details
Gene Symbol: LINC01088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038342.1 | 6 | 551 | |||
GenBank mRNA | BC036848.1 | - | 6 | 551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI914854.1 | - | 6 | 551 | Chr. 4 - 78971753 - 79308429 [+] on Build GRCh38 |
