siRNA Details
Gene Symbol: LINC01091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027106.1 | 3 | 1938 | |||
GenBank mRNA | BC033378.1 | - | 3 | 1938 | ||
BC046914.1 | - | 3 | 1938 | |||
BC053945.1 | - | 3 | 1938 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027106.1 | 3 | 1938 | |||
GenBank mRNA | BC033378.1 | - | 3 | 1938 | ||
BC046914.1 | - | 3 | 1938 | |||
BC053945.1 | - | 3 | 1938 |