siRNA Details
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126481.1 | - | 5 | 1651 | ||
AL137734.1 | - | 1 | 1990 | |||
CR933654.1 | - | 2 | 4615 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126481.1 | - | 5 | 1651 | ||
AL137734.1 | - | 1 | 1990 | |||
CR933654.1 | - | 2 | 4615 |