siRNA Details
Gene Symbol: NOP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015453.2 | 4 | 2986 | |||
GenBank mRNA | AB000464.1 | - | 4 | 2986 | ||
AB000465.1 | - | 3 | 868 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NOP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015453.2 | 4 | 2986 | |||
GenBank mRNA | AB000464.1 | - | 4 | 2986 | ||
AB000465.1 | - | 3 | 868 |