siRNA Details
Gene Symbol: NOP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015453.2 | 1 | 166 | |||
GenBank mRNA | AB000464.1 | - | 1 | 166 | ||
AB000466.1 | - | 1 | 166 | |||
AK054619.1 | - | 1 | 813 | |||
BC020978.1 | - | 1 | 788 | |||
BC032331.1 | - | 1 | 772 |
