siRNA Details
Gene Symbol: SRD5A3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037969.1 | 4 | 458 | |||
GenBank mRNA | BC034006.1 | - | 4 | 463 | ||
BC047403.1 | - | 3 | 365 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SRD5A3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037969.1 | 4 | 458 | |||
GenBank mRNA | BC034006.1 | - | 4 | 463 | ||
BC047403.1 | - | 3 | 365 |