siRNA Details
Gene Symbol: LOC101928174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120329.1 | 2 | 290 | |||
GenBank mRNA | AK092087.1 | - | 2 | 290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA685006.1 | - | 2 | 292 | Chr. 15 - 34755084 - 34810734 [+] on Build GRCh38 |
