siRNA Details
Gene Symbol: LINC01018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024423.2 | 5 | 1380 | |||
NR_024424.2 | 3 | 1394 | ||||
GenBank mRNA | AK125283.1 | - | 5 | 1347 | ||
BC043253.2 | - | 3 | 1083 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024423.2 | 5 | 1380 | |||
NR_024424.2 | 3 | 1394 | ||||
GenBank mRNA | AK125283.1 | - | 5 | 1347 | ||
BC043253.2 | - | 3 | 1083 |