siRNA Details
Gene Symbol: RDH10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125388.1 | 2, 3 | 247 | |||
GenBank mRNA | AK128216.1 | - | 2, 3 | 247 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RDH10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125388.1 | 2, 3 | 247 | |||
GenBank mRNA | AK128216.1 | - | 2, 3 | 247 |