siRNA Details
Gene Symbol: LOC101928203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110934.1 | 3 | 326 | |||
GenBank mRNA | AK057218.1 | - | 3 | 326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB019544.1 | - | 3 | 326 | Chr. 16 - 76634998 - 76657097 [+] on Build GRCh38 |
