siRNA Details
Gene Symbol: LOC105374631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134266.1 | 1 | 50 | |||
GenBank mRNA | BX089348.1 | - | 1 | 50 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105374631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134266.1 | 1 | 50 | |||
GenBank mRNA | BX089348.1 | - | 1 | 50 |