siRNA Details
Gene Symbol: LNX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046622.1 | 3 | 248 | |||
GenBank mRNA | AA405946.1 | - | 3 | 248 | ||
BX103714.1 | - | 3 | 247 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA496074.1 | - | 3 | 248 | Chr. 4 - 53500078 - 53523164 [+] on Build GRCh38 |
