siRNA Details
Gene Symbol: LOC101928622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125902.1 | 5 | 1949 | |||
GenBank mRNA | BC036345.1 | - | 5 | 1950 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125902.1 | 5 | 1949 | |||
GenBank mRNA | BC036345.1 | - | 5 | 1950 |