siRNA Details
Gene Symbol: LOC102467213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104671.1 | 4 | 374 | |||
GenBank mRNA | AA292843.1 | - | 4 | 374 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY021255.1 | - | 4 | 362 | Chr. 5 - 106815291 - 107011002 [-] on Build GRCh38 |
