siRNA Details
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033900.1 | 3, 4 | 1074 | |||
GenBank mRNA | BC034307.1 | - | 3, 4 | 1015 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB507707.2 | - | 1, 2 | 508 | Chr. 4 - 186286098 - 186288926 [-] on Build GRCh38 |
