siRNA Details
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033900.1 | 3 | 692 | |||
GenBank mRNA | BC034307.1 | - | 3 | 633 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033900.1 | 3 | 692 | |||
GenBank mRNA | BC034307.1 | - | 3 | 633 |