siRNA Details
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033900.1 | 3 | 515 | |||
GenBank mRNA | BC034307.1 | - | 3 | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG719069.1 | - | 3 | 461 | Chr. 4 - 186288937 - 186500999 [-] on Build GRCh38 |
