siRNA Details
Gene Symbol: LINC00967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039979.1 | 1 | 124 | |||
GenBank mRNA | BX360434.2 | - | 1 | 128 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039979.1 | 1 | 124 | |||
GenBank mRNA | BX360434.2 | - | 1 | 128 |