siRNA Details
Gene Symbol: HOXA10-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046609.1 | 2 | 340 | |||
GenBank mRNA | BM549446.1 | - | 2 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ923616.1 | - | 1 | 490 | Chr. 7 - 27169072 - 27171505 [+] on Build GRCh38 |
