siRNA Details
Gene Symbol: LINC00293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027012.1 | 9 | 715 | |||
NR_027013.1 | 8 | 1262 | ||||
GenBank mRNA | AK097475.1 | - | 9 | 715 | ||
BC041354.1 | - | 8 | 1274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR739932.1 | - | 7 | 576 | Chr. 8 - 46840895 - 46854211 [+] on Build GRCh38 |
