siRNA Details
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051989.1 | 4 | 821 | |||
GenBank mRNA | BG494657.1 | - | 4 | 557 | ||
BG496346.1 | - | 3 | 657 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051989.1 | 4 | 821 | |||
GenBank mRNA | BG494657.1 | - | 4 | 557 | ||
BG496346.1 | - | 3 | 657 |